Nalgene® Rectangular Bottles, HDPE, Bulk Pack, Thermo Scientific
À propos de cet article
Nalgene® high density polyethylene rectangular bottles come with polypropylene closures.
Bottles are available in both translucent and amber. Translucence allows easy viewing of liquid levels. Amber reduces UV light transmissions to protect light-sensitive liquids.
All the bottles are leak proof, and feature a space-saving rectangular design. These bottles feature uniform walls which are resistant to splitting or puncturing.
: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
: Polypropylene screw closures are included.
: They are bulk packed for packaging applications.
7 Options Available Below
Product Details & Documents
Nalgene® high density polyethylene rectangular bottles come with polypropylene closures.
Bottles are available in both translucent and amber. Translucence allows easy viewing of liquid levels. Amber reduces UV light transmissions to protect light-sensitive liquids.
All the bottles are leak proof, and feature a space-saving rectangular design. These bottles feature uniform walls which are resistant to splitting or puncturing.
: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
: Polypropylene screw closures are included.
: They are bulk packed for packaging applications.
Product Family Options
Natural
Informations sur le produit
- CapacitéHeight with CapNeck I.D.Taille du bouchonAvailabilityPrice
- 125 ml (4.2 oz.)102 mm21 mm28 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 500Case of 500 - 1 464,00 $1 464,00 $Prix promotionnel pour les nouveaux clientsCase of 5001 464,00 $Supplier Number: 312007-0004
- 250 ml (8.5 oz.)117 mm28 mm38 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 250Case of 250 - 944,00 $944,00 $Prix promotionnel pour les nouveaux clientsCase of 250944,00 $Supplier Number: 312007-0008
- 500 ml (16.9 oz.)147 mm37 mm48 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 125Case of 125 - 799,20 $799,20 $Prix promotionnel pour les nouveaux clientsCase of 125799,20 $Supplier Number: 312007-0016
- 1 L (33.8 oz.)180 mm44 mm53 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 50Case of 50 - 680,80 $680,80 $Prix promotionnel pour les nouveaux clientsCase of 50680,80 $Supplier Number: 312007-0032
Amber
Informations sur le produit
- CapacitéHeight with CapNeck I.D.Taille du bouchonAvailabilityPrice
- 125 ml (4.2 oz.)102 mm21 mm28-415Log in pour la disponibilitéLog in pour la disponibilitéCase of 500Case of 500 - 2 158,60 $2 158,60 $Prix promotionnel pour les nouveaux clientsCase of 5002 158,60 $Supplier Number: 312009-0004
- 250 ml (8.5 oz.)117 mm28 mm38-415Log in pour la disponibilitéLog in pour la disponibilitéCase of 250Case of 250 - 1 688,00 $1 688,00 $Prix promotionnel pour les nouveaux clientsCase of 2501 688,00 $Supplier Number: 312009-0008
- 1 L (33.8 oz.)180 mm44 mm48-415Log in pour la disponibilitéLog in pour la disponibilitéCase of 50Case of 50 - 954,60 $954,60 $Prix promotionnel pour les nouveaux clientsCase of 50954,60 $Supplier Number: 312009-0032