Closures, Polyethylene, Greiner Bio-One
Greiner Bio-One closures are made of high-grade polyethylene and are available in many colors.
- High-grade polyethylene construction
- Available in a variety of colors
- Two options for closure type
There are two different types of closures: Grip Stoppers and Screw caps.
Frequently Bought Together