Nalgene® Graduated Square Bottles, PPCO, Bulk Pack, Thermo Scientific
Nalgene® graduated polypropylene copolymer square bottles are autoclavable, and have molded-in graduations for easy measuring.
The bottles feature a square design which helps save space. These bottles have a drip-proof neck finish for convenient dispensing.
: Bottles are packaged in bulk. The number of bottles in the package depends on the size of the bottle.
Frequently Bought Together