Ion Selective Electrodes, Hach | Avantor
Ion Selective Electrodes, Hach
Supplier:   Hach
Ion Selective Electrodes, Hach
Supplier:  Hach
Certificates
Ion Selective Electrodes, Hach
1 Options Available Below
Product Details & Documents
Hach offers a wide range of electrodes for every application and budget
Our wide selection of metal electrodes, combined metal electrodes, and ion selective electrodes will have the right probe for your application.
Certifications: .CE.WEEE.
Ordering information: The sensION+ electrodes require a reference electrode for use. Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89501-156
  • CA89501-156
  • Connection:
  • BNC
  • BNC
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Plage de température
    Availability
    Price
  • sensION+ ISE Electrode, Nitrate, sensor only, no reference
    0...50 °C
    Discontinued
    Supplier Number: -
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89501-156
    Connection
    BNC
    Product available on GSA Advantage®
  • sensION+ ISE Electrode, Nitrate, sensor only, no reference
    0...50 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 2 289,23 $
    2 289,23 $
    Each
       2 289,23 $
    Supplier Number: LZW9662C.97.002
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89501-156
    Connection
    BNC
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Chloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette Ampules
    Chloride Standard Solutions as Cl-(NIST) 12,500 mg/L, 16/Voluette Ampules
    CA89173-990
    Log in pour la disponibilité
    Pack of 16 (10ml) - $258.43