ISE Electrodes for HQd Meter, Hach
ISE Electrodes for HQd Meter, Hach
Supplier:   Hach
ISE Electrodes for HQd Meter, Hach
Supplier:  Hach
Certificates
ISE Electrodes for HQd Meter, Hach
ISE Electrodes for HQd Meter, Hach
4 Options Available Below
Product Details & Documents
Hach®, the leader in water quality, has developed a customized electrochemistry solution for water quality testing that takes the guesswork out of your measurements
Factory-calibrated IntelliCAL probes are ready for use, right out of the box. Each Hach® IntelliCAL probe stores its calibration history and settings in the probe itself, allowing the probes to easily be transferred from meter to meter, minimizing probe replacement and calibration. The serial number is also stored in the probe. Probes issue alerts when re-calibration is needed, but can be moved between meters without re-calibration.
Designed for your water applications, the Hach® HQd smart probes automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time.
Ordering information: Many ISE electrodes require standards, filling solutions, ISA, etc. for electrode use. Be sure to check these requirements when ordering your Hach ISE electrode.
Product Family Options
Product Information
  • Désignation
    Matière
    For Use With
    Longueur
    Cable Length
    Connecteur
    Fill Solution
    Measurement Range
    Plage de température
    Junction
    Availability
    Price
  • IntelliCAL Ammonia ISE Electrode
    Epoxy
    HQ30d, HQ40d, HQ430d, HQ440
    220 mm
    1 m
    HQD specific
    Ammonia, 50 ml
    0.007 mg/L - 14,000 mg/L
    0...50 °C
    Porous PTFE annular ring
    Log in for availability
    Log in for availability
    Each
    Each - $2,883.37
    $2,883.37
    Each
       $2,883.37
    Supplier Number: ISENH318101
  • IntelliCAL Fluoride ISE Electrode
    Epoxy
    HQ30d, HQ40d, HQ430d, HQ440
    220 mm
    1 m
    HQD specific
    Non-refillable driTEK ge
    0.02 mg/L - 19,000 mg/L
    5...50 °C
    Porous PTFE annular ring
    Log in for availability
    Log in for availability
    Each
    Each - $3,394.89
    $3,394.89
    Each
       $3,394.89
    Supplier Number: ISEF12101
  • IntelliCAL ISENa381 Sodium Ion Selective Electrode (ISE)
    Zeonor™
    Multiple HQd Series Laboratory and Portable Meters
    220 mm
    3 m
    HQD specific
    NH₄Cl, 0.02 M
    0.023 mg/L - 23,000 mg/L
    0...50 °C
    Porous PTFE annular ring
    Discontinued
    Supplier Number: -
  • IntelliCAL ISENa381 Sodium Ion Selective Electrode (ISE)
    Zeonor™
    Multiple HQd Series Laboratory and Portable Meters
    220 mm
    3 m
    HQD specific
    NH₄Cl, 0.02 M
    0.023 mg/L - 23,000 mg/L
    0...50 °C
    Porous PTFE annular ring
    Log in for availability
    Log in for availability
    Each
    Each - $1,877.59
    $1,877.59
    Each
       $1,877.59
    Supplier Number: ISENA38103
  • IntelliCAL ISENa381 Sodium Ion Selective Electrode (ISE)
    Zeonor™
    HQ30d, HQ40d, HQ430d, HQ440
    220 mm
    1 m
    HQD specific
    NH₄Cl, 0.02 M
    0.023 mg/L - 23,000 mg/L
    0...50 °C
    Ceramic pin
    Log in for availability
    Log in for availability
    Each
    Each - $2,421.85
    $2,421.85
    Each
       $2,421.85
    Supplier Number: ISENA38101
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Ammonia Standard Solution, NH3-N 150 mg/L, 16 - 10 mL Voluette Ampule for TKN
    Ammonia Standard Solution, NH3-N 150 mg/L, 16 - 10 mL Voluette Ampule for TKN
    CA89173-996
    Log in for availability
    Pack of 16 - $258.62
Product Information
  • Catalog #
    Availability
    Price
  • Ammonium Ionic Strength Adjustor Powder Pillows (IntelliCAL only)
    Ammonium Ionic Strength Adjustor Powder Pillows (IntelliCAL only)
    CA89173-994
    Log in for availability
    Each - $236.53
Product Information
  • Catalog #
    Certification
    Availability
    Price
  • IntelliCAL ISENH3181 Replacement Membrane Modules, 3/pk
    IntelliCAL ISENH3181 Replacement Membrane Modules, 3/pk
    CA89174-118
    RoHS
    Log in for availability
    Each - $406.77
Product Information
  • Catalog #
    Availability
    Price
  • NITRATE IONIC STRG ADJ PP, PK/100
    NITRATE IONIC STRG ADJ PP, PK/100
    CA89173-992
    Log in for availability
    Pack of 100 - $228.10