About this item
Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer
Blue- kerosene, Yellow - diesel, Green-oils.
Protection features include a 3-1/2" (88mm) long chemical resistant stainless steel flame arrester that stops flashback ignition, provides faster liquid flow, and exceptional durability. The self-venting, self-closing leakproof lid prevents ruptures from pressure build-up and controls vapors and spills. Warning label includes an extra large ID zone for user labeling of contents to avoid misuse, reduce the potential for incompatible mixtures, or may be used to designate department or individual owner. Made of galvanized steel with a tough powder paint finish, can capacities 3.8L (1gal.) and above have ample headspace to accommodate oil additives.