Non-Ionic Detergent Solutions
Non-Ionic Detergent Solutions
Non-Ionic Detergent Solutions
Supplier:  G-Biosciences
Certificates
Non-Ionic Detergent Solutions
22 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Non-ionic detergents have a hydrophilic head group that is uncharged and are preferred for their ability to break lipid-lipid and lipid-protein interactions.
The non-ionic deteregents are supplied as a general Research Grade, Proteomic Grade (PG) Solutions and 2D-Detergents™. The Proteomic Grade (PG) Solutions have ultra low aldehyde (<100 μM) and peroxide (<50 μM) concentrations to reduce the effects of peroxidase and carbonyl compounds that negatively interact with membrane proteins. The 2D-Detergents™ have low conductivity (<10 μS) and ultra low aldehyde (<100 μM) and peroxide (<50 μM) concentrations.
G-Biosciences Non-Ionic Detergent Solutions include: Triton® X-100, Triton® X-114, Brij® 35, Brij® 58, Tween® 20, Tween® 80, Nonidet® P-40 Substitute, Octyl ß Glucoside, MEGA 8, MEGA 9, MEGA 10, BigCHAP, and Deoxy Big CHAP.
They have limited ability to break protein-protein interactions and are often referred to as non-denaturing detergents and are used to isolate biologically active membrane proteins.
Ordering information: Accessories not included.
Documents
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • Octyl ß Glucoside
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $667.15
    $667.15
    Each (1g)
       $667.15
    Supplier Number: DG015
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Octyl ß Glucoside
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $2,922.52
    $2,922.52
    Each (5g)
       $2,922.52
    Supplier Number: DG016
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 8
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $701.97
    $701.97
    Each (1g)
       $701.97
    Supplier Number: DG017
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 8
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $2,683.54
    $2,683.54
    Each (5g)
       $2,683.54
    Supplier Number: DG018
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 9
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $701.97
    $701.97
    Each (1g)
       $701.97
    Supplier Number: DG019
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 9
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $2,414.60
    $2,414.60
    Each (5g)
       $2,414.60
    Supplier Number: DG020
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 10
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $712.13
    $712.13
    Each (1g)
       $712.13
    Supplier Number: DG021
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • MEGA 10
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $2,683.54
    $2,683.54
    Each (5g)
       $2,683.54
    Supplier Number: DG022
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Big CHAP
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $975.86
    $975.86
    Each (1g)
       $975.86
    Supplier Number: DG023
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Big CHAP
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $3,838.48
    $3,838.48
    Each (5g)
       $3,838.48
    Supplier Number: DG024
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Deoxy Big CHAP
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $2,151.92
    $2,151.92
    Each (1g)
       $2,151.92
    Supplier Number: DG025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Deoxy Big CHAP
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $8,065.41
    $8,065.41
    Each (5g)
       $8,065.41
    Supplier Number: DG026
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Tween® 80
    500 ml
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $291.57
    $291.57
    Each (1l)
       $291.57
    Supplier Number: 786-519
  • Tween® 80
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $489.65
    $489.65
    Each (1l)
       $489.65
    Supplier Number: 786-520
  • Brij® 35
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $686.06
    $686.06
    Each (500g)
       $686.06
    Supplier Number: 786-521
  • Brij® 58
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $767.92
    $767.92
    Each (500g)
       $767.92
    Supplier Number: 786-522
  • Nonidet® P-40 Substitute
    500 ml
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $829.40
    $829.40
    Each (1l)
       $829.40
    Minimum order quantity is 1
    Supplier Number: 786-511
  • Nonidet® P-40 Substitute
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $1,622.32
    $1,622.32
    Each (1l)
       $1,622.32
    Minimum order quantity is 1
    Supplier Number: 786-512
  • Tween® 20
    500 ml
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $209.51
    $209.51
    Each (1l)
       $209.51
    Minimum order quantity is 1
    Supplier Number: 786-517
  • Tween® 20
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $375.95
    $375.95
    Each (1l)
       $375.95
    Minimum order quantity is 1
    Supplier Number: 786-518
  • Brij® 35
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $278.95
    $278.95
    Each (250g)
       $278.95
    Supplier Number: 786-351
  • Brij® 58
    250 g
    Log in for availability
    Log in for availability
    Each (250g)
    Each (250g) - $322.16
    $322.16
    Each (250g)
       $322.16
    Supplier Number: 786-352