ContactUsLinkComponent
Protein Concentrator Solution, G-Biosciences
Protein Concentrator Solution, G-Biosciences
Catalog # CA89167-726
Supplier:  G-Biosciences
CAS Number:  
Protein Concentrator Solution, G-Biosciences
Catalog # CA89167-726
Supplier:  G-Biosciences
Supplier Number:  786-143
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Concentrator Solution
  • Cat. no.:
    CA89167-726
  • Supplier No.:
    786-143
Specifications
About this item
Concentrator Solution is a novel liquid polymer for the rapid concentration of dilute protein solutions with zero loss, using dialysis
The water will be rapidly removed through the dialysis membrane, which also retains your protein of interest and prevents the high molecular weight liquid polymer entering your solution. Once the desired volume of your solution is achieved, quickly rinse the excess concentrator solution from the dialysis bag/ membrane and recover your sample.
Concentrator Solution is also supplied with our Tube-O-DIALYZER™ as Tube-O-CONCENTRATOR™. Use molecular weight cut offs (MWCO) of 30,000 daltons or less to avoid cross contamination.
Simply transfer your dilute protein solution to a dialysis bag or dialysis device, such as our patented Tube-O-DIALYZER™ and dialyze against the concentrator solution.
Ordering information: Accessories not included.