ContactUsLinkComponent
SG-Chymotrypsin™
Catalogue # CA82022-518
Fournisseur:  G-Biosciences
Numéro CAS:  
SG-Chymotrypsin™
Catalogue # CA82022-518
Fournisseur:  G-Biosciences
Supplier Number:  786-13
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    2×25 μg
  • Enzyme name:
    SG-Chymotrypsin™
  • Grade:
    Sequencing Grade
  • Cat. no.:
    CA82022-518
  • Supplier No.:
    786-13
Spécifications
À propos de cet article
SG-Chymotrypsin™ is a serine endopeptidase, which predominantly cleaves peptide bonds on the carboxy side of tyrosine, phenylalanine and tryptophan
SG-Chymotrypsin™ is a modified chymotrypsin with high specificity, free from other known endopeptidases, that is resistant to autolysis and degradation. SG-Chymotrypsin™ is used for the digestion of proteins for sequence analysis and is suitable for sequencing applications. Use of SG-Chymotrypsin™ produces consistent activity levels from lot to lot for reproducible digestions.
SG-Chymotrypsin™ is first treated with TLCK to inhibit trypsin that may be present, and then subjected to an extensive purification process to remove contaminating protease and chymotryptic autolysis by-products. The highly purified enzyme is then chemically modified to increase its resistance to autolysis and stability. For protein digestion, SG-Chymotrypsin™ is added to the protein at a ratio of 1:200 to 1:50, by weight, in a standard digestion buffer. Incubate at 25-30°C for 1 to 10 hours, but can be extended to 24 hours, due to the extended life of the SG-Chymotrypsin™. We recommend choosing a ratio of enzyme to protein that allows for the shortest incubation time possible. This will reduce or eliminate the catalyzed hydrolysis of peptide bonds with non-aromatic amino acid residues.
In addition, chymotrypsin has a low catalytic activity against the carboxy side of leucine, methionine, alanine, aspartic and glutamic acids. It is therefore recommended to always use the shortest digestion time possible.