BIOPURE 15 Water Purification System with Booster Pump
CA89204-162
: Avantor
- Description:BIOPURE 15 with booster pump
- Cat. no.:CA89204-162
- Supplier No.:MB015BPM1
The Biopure 15 is ideal for ENT and dental decontamination.
- Dispense Rate up to 2.0 LPM
- Make Up Flow Rate 5 LPH (360 L/day)
- Water Quality ype II
High Performance Water Purification System designed to feed washer disinfectors and endoscope reprocessors. Feed water booster pump included.
It can directly supply 2 L/min of compliant purified water directly to a wide range of applications including washer disinfectors, autoclaves, Ultra sonic baths, ENT Automated Endoscope Reprocessors, etc. The Biopure 15 can produce up to 15 L/hr of purified water.