ELGA Deionization Systems
Supplier:   John Meunier
ELGA Deionization Systems
Supplier:  John Meunier
Certificates
ELGA Deionization Systems
ELGA Deionization Systems
2 Options Available Below
Product Details & Documents
ELGA’s range of wall mounted deionization systems produce from 30 to 90 liters per hour of high purity water for a wide range of laboratory and general manufacturing applications.
  • Cost-effective
  • Easy monitoring
  • Built-in meters
  • Water Quality: Type II
The deionizers are cost-effective stand-alone systems that provide low to medium volumes of primary or deionized grade water. Water quality is easy to monitor, with built in meters or colour change indicators.
Product Family Options
Informations sur le produit
  • Désignation
    W×D×H
    Débit
    Weight
    Availability
    Price
  • B114 deionizer
    180×152×380 mm (7.1×6×15")
    30 LPH
    2.2 kg (4.9 lbs.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,380.82
    $1,380.82
    Each
       $1,380.82
    Supplier Number: B114/A
  • Vision 125 deionizer
    92×110×385 mm (3.6×4.3×15.2")
    60 LPH
    2.5 kg (5.5 lbs.)
    Discontinued
    Supplier Number: -
  • Vision 250 deionizer
    92×110×660 mm (3.6×4.3×26")
    60 LPH
    4.2 kg (9.3 lbs.)
    Discontinued
    Supplier Number: -
  • Micromeg deionizer
    200×230×550 mm (7.9×9×21.7")
    90 LPH
    6.5 kg (14.3 lbs.)
    Discontinued
    Supplier Number: -
Deionizer consumables
Informations sur le produit
  • Désignation
    W×D×H
    Débit
    Weight
    Availability
    Price
  • Deionizer replacement cartridge, for Vision 125, filter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $373.65
    $373.65
    Each
       $373.65
    Supplier Number: VC125