Tank Tool, Micronova
This unique tool features a malleable plate that flexes to create a custom curve to match a range of tanks and cylinder shapes in clean and controlled environments. Available in interior or exterior configurations.
- Flexible mop head for custom shaping
- Easily clean and sanitize tanks and cylinders
- Available with a plastic or stainless steel plate
- SnapMop™ and PocketMop™ series covers available in a range of fabrics
- For use in clean and controlled environments
- Compatible with strong disinfectants
- Autoclavable
The Micronova TankTool™ is designed to be used with SnapMop™ and PocketMop™ series covers. It can be used with SSU series universal handles or STA series threaded handles for extended reach.
All components are ordered separately. For additional handle lengths, see Universal Mop Handles or Plastic Mop Handles from Micronova. See SnapMop™ and PocketMop™ for additional cover options.
: Accessories not included.
Handles
Frequently Bought Together