QuickTask™ Flat Mops, Contec®
QuickTask™ Flat Mops, Contec®
QuickTask™ Flat Mops, Contec®
Supplier:  CONTEC, INC. MX
Certificates
QuickTask™ Flat Mops, Contec®
QuickTask™ Flat Mops, Contec®
8 Options Available Below
Product Details & Documents
The QuickTask™ product line consists of a variety of knitted polyester, polyester/cellulose, microfiber/foam, and microfiber flat mop heads that are easily installed onto a hinged mop head frame and handle assembly.
  • Mop heads available in various fabrics
  • Mop head covers are compatible with a wide range of chemicals and disinfectants
  • Available in sterile and non-sterile formats
  • QuickTask™ hardware is fully autoclavable
Each mop head has been specifically chosen to be compatible with a wide range of chemicals and disinfectants including quats and hypochlorite solutions. QuickTask mop head frames are available in stainless steel or durable autoclavable plastic.
Model TASK0200 and TASK0250 (Sterile): The lightest mop cover in the range, the sorbent cleanroom laundered multi-layer knitted polyester is ideal for use on smooth surfaces. Its sorbent knit structure applies solutions evenly and removes contaminants uniformly. Available validated sterile by gamma irradiation.
Model TASK0300: The most sorbent mop cover in the range, the small, blue loop microfiber construction of TASK0300 provides exceptional performance in removing liquids, soil, residues, and other contaminants from most surfaces. It has exceptional sorbency and mop-to-dry performance on wet surfaces. It is compatible with chlorine solutions and effectively cleans polymeric flooring.
Model TASK0400: The cleanest and most durable mop cover in the range, TASK0400 has tubular knit loops that conform to almost any shape, making cleaning of cove bases and ceiling and wall joints easier than with standard flat mops. Its microfiber construction provides exceptional sorbency and mop-to-dry performance on wet surfaces. It is compatible with a wide range of chemicals and disinfectants, including chlorine solutions.
Model TASK0500, TASK0550 (Irradiated): The non-woven mop head in the range features a polyester/cellulose cover over a polyester fiberfill core. It is ideal for cost effective single use. Its non-woven, highly sorbent fabric construction allows for removal of light soils, residues, and biofilms. Available gamma irradiated.
Model TASK0600: The unique microfiber face and foam core construction ensures excellent floor contact, sorbency, and superior surface cleaning. Durable and economical, allowing for cost effective single use.
Product Family Options
Product Information
  • Désignation
    Dimensions
    Matière
    Sterility
    Color
    Packed
    Availability
    Price
  • QuickTask™ Magnetic mop frame
    40×10 cm (15.5×3.75")
    Stainless steel
    Non sterile
    Grey
    1/case
    Log in for availability
    Log in for availability
    Each
    Each - $528.90
    $528.90
    New Customer Promotional Price
    Each
       $528.90
    Supplier Number: 2906
  • QuickTask™ Magnetic mop frame
    40×10 cm (15.5×3.75")
    PP
    Non sterile
    Blue
    1/case
    Log in for availability
    Log in for availability
    Each
    Each - $210.70
    $210.70
    New Customer Promotional Price
    Each
       $210.70
    Supplier Number: 2699P
  • QuickTask™ Polyester Knitted Mop Head
    41×12.7 cm (16×5")
    Polyester knit
    Non sterile
    White
    4/Bag, 12 Bags/Case
    Log in for availability
    Log in for availability
    Case of 48
    Case of 48 - $681.12
    $681.12
    New Customer Promotional Price
    Case of 48
       $681.12
    Supplier Number: TASK0200
  • QuickTask™ Sterile Polyester Knitted Mop Head
    41×12.7 cm (16×5")
    Polyester knit
    Sterile
    White
    1/Bag, 40 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $937.40
    $937.40
    New Customer Promotional Price
    Case of 40
       $937.40
    Supplier Number: TASK0250
  • QuickTask™ Microfiber Mop Head
    41×12.7 cm (16×5")
    Microfiber, looped
    Non sterile
    Blue
    10/Bag, 4 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $656.18
    $656.18
    New Customer Promotional Price
    Case of 40
       $656.18
    Supplier Number: TASK0300
  • QuickTask™ Polyester Knitted Microfiber Mop Head
    41×12.7 cm (16×5")
    Knitted looped polyester microfiber
    Non sterile
    White
    10/Bag, 5 Bags/Case
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $1,419.00
    $1,419.00
    New Customer Promotional Price
    Case of 50
       $1,419.00
    Supplier Number: TASK0400
  • QuickTask™ Polyester/Cellulose Mop Head
    41×12.7 cm (16×5")
    Polyester/cellulose
    Non sterile
    White
    4/Bag, 12 Bags/Case
    Discontinued
    Supplier Number: TASK0500
  • QuickTask™ Irradiated Polyester/Cellulose Mop Head
    41×12.7 cm (16×5")
    Polyester/cellulose
    Sterile
    White
    1/Bag, 40 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $322.50
    $322.50
    New Customer Promotional Price
    Case of 40
       $322.50
    Supplier Number: TASK0550
  • QuickTask™ Polyester Microfiber/Foam Mop Head
    41×12.7 cm (16×5")
    Polyester microfiber/foam
    Non sterile
    White
    1/Bag, 40 Bags/Case
    Log in for availability
    Log in for availability
    Case of 40
    Case of 40 - $1,135.20
    $1,135.20
    New Customer Promotional Price
    Case of 40
       $1,135.20
    Supplier Number: TASK0600