Corning® HYPERStack™ Stainless Steel Manipulator, Corning
Corning® HYPERStack® Stainless Steel Manipulator
Catalog # 89184-638
Supplier: Corning
Fournisseur #: 11000
Jump to:
Product Details & Documents
The Corning® HYPERStack™ Stainless Steel Manipulator accommodates both HYPERStack™-36 and CellSTACK-10 vessels.
It handles two units of the same type simultaneously or one unit at a time. It is ergonomically designed and has large, easy to read dials that are clearly marked to identify positions for ease of use in protocol development.
Spécifications
- Désignation:Corning® HYPERStack® Stainless Steel Manipulator