Semi-quantitative test strips QUANTOFIX Peroxide 100
100 Sticks
Supplier: Macherey-Nagel
Fournisseur #: 91312
Semi-quantitative test strips QUANTOFIX Peroxide 100
Catalog # 60787-736
Supplier: Macherey-Nagel
CAS Number:
Jump to:
Product Details & Documents
For semi-quantitative determinations
Gradation: 0 to 100mg/L. Color changes from white to blue.
Documents
Certificates
Recherche par numéro de lotSpécifications
Spécifications
- Type de conditionnement:100 Sticks