Anti-TYMS Rabbit Polyclonal Antibody
CA100-401-199
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:Thymidylate Synthase
- Antigen symbol:TYMS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µL
- Cross adsorption:No
- Format:Antiserum
- Form:Liquid (sterile filtered)
- Gene ID:7298
- Antigen synonyms:TS|d TMP synthase|EC 2.1.1.45|TMS|MGC88736|HsT422
- Storage buffer:0.01% (w/v) Sodium Azide,
- Molecular weight:36 kDa
- Storage temperature:–20 °C
- Concentration:85 mg/ml by Refractometry
- Shipping temperature:Dry Ice
- Immunogen:This whole rabbit serum was prepared by repeated immunizations with recombinant human Thymidylate Synthase (36 kDa) produced in E.coli.
- Cat. no.:CA100-401-199
- Supplier No.:100-401-199
This antiserum against Human Thymidylate Synthase (hTS) has been tested for use in ELISA, immunoprecipitation, immunofluorescence microscopy, immunochemistry and immunoblotting.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: TYMS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human