To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:GLI Family Zinc Finger 2
- Antigen symbol:GLI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:2736
- Antigen synonyms:THP2|HPE9|CJS|PHS2|THP1
- Amino acid number:46 to 60
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 46-60 of human Gli-2 (isoform a).
- Purification:affinity-purified
- Cat. no.:CA600-401-845
- Supplier No.:600-401-845
Specifications
About this item
This antibody has been tested for use in ELISA, immunochemistry and western blot. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:2000 - 1:12000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:500 - 1:2000;
Type: Primary
Antigen: GLI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human