Nalgene® Polypropylene Spigot Closure, Thermo Scientific
Catalog # 27802-069
Supplier: Thermo Fisher Scientific
: 6423-0010
Nalgene® Polypropylene Spigot Closure, Thermo Scientific
Catalog # 27802-069
Supplier: Thermo Fisher Scientific
Product Details & Documents
Nalgene® polypropylene spigot closures can be used to replace spigots to provide a leakproof seal when autoclaving liquids in a carboy.
They are also useful when handling could damage or accidentally turn on spigot, or when spigot-less carboy is required. These spigot closures fit all Nalgene® carboys and selected Nalgene® tanks with spigots.
Ordering information: Included with all NALGENE® carboys with spigot.