Corning® HYPERStack® Cell Culture Vessels
Corning® HYPERStack® Cell Culture Vessels
Supplier:   Corning
Corning® HYPERStack® Cell Culture Vessels
Supplier:  Corning
Certificates
Corning® HYPERStack® Cell Culture Vessels
Corning® HYPERStack® Cell Culture Vessels
8 Options Available Below
Product Details & Documents
Corning® HYPERStack® cell culture vessels combine the best of two Corning products: the Corning CellSTACK® and the Corning HYPERFlask® cell culture vessels. The utilization of gas-permeable film technology provided in the spatial footprint of the CellSTACK vessel allows the HYPERStack platform to be the most efficient, scalable cell culture vessels for adherent cell culture available today.
  • More cells - provides up to 5X the growth surface area of a traditional cell culture vessel of comparable footprint
  • Closed system - no open fluid manipulations
  • Scalable product - multiple size offerings support scale-up and scale-out
  • Ergonomic design - easier manipulation with handling equipment and accessories
  • Fixed media volume - 0.2 ml/cm² fills vessel for less volumetric waste
  • Sterile - sterility assurance level (SAL) of 10⁻⁶
For accessories designed to facilitate the use of Corning HYPERStack cell culture vessels please see:
Corning 2 L Erlenmeyer flask with aseptic transfer cap (Cat. No. 89204-710), Corning 5 L Erlenmeyer flask with aseptic transfer cap (Cat. No. 10790-810), Corning Disposable Tubing Set for 850 cm² polystyrene bottle (89184-632), Corning 850 cm² polystyrene bottle (89184-640).
Product Family Options
Single vent filters
Product Information
  • Désignation
    Working Volume
    Growth Area
    Availability
    Price
  • HYPERStack® - 36 layer cell culture vessels, Corning® CellBIND® surface
    3900 ml
    18000 cm² (500 cm² per layer)
    Discontinued
    Supplier Number: -
  • HYPERStack® - 12 layer cell culture vessels, Corning® CellBIND® surface
    1310 ml
    6000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $4,380.00
    $4,380.00
    Case of 4
       $4,380.00
    Supplier Number: 20012
  • HYPERStack® - 12 layer cell culture vessels, non-treated surface
    1310 ml
    6000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $4,590.00
    $4,590.00
    Case of 4
       $4,590.00
    Supplier Number: 20013
  • HYPERStack® - 36 layer cell culture vessels, Corning® CellBIND® surface
    3920 ml
    18000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $4,640.00
    $4,640.00
    Case of 2
       $4,640.00
    Supplier Number: 20036
  • HYPERStack® - 36 layer cell culture vessels, non-treated surface
    3920 ml
    18000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $4,500.00
    $4,500.00
    Case of 2
       $4,500.00
    Supplier Number: 20037
Dual vent filters
Product Information
  • Désignation
    Working Volume
    Growth Area
    Availability
    Price
  • HYPERStack® - 12 layer cell culture vessels, dual vent filter, Corning® CellBIND® surface
    1310 ml
    6000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $11,290.00
    $11,290.00
    Log in for availability
    Each
    Each - $11,290.00
    $11,290.00
    Case of 4
       $11,290.00
    Each
       $11,290.00
    Supplier Number: 24012
  • HYPERStack® - 12 layer cell culture vessels, dual vent filter, non-treated surface
    1310 ml
    6000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $11,290.00
    $11,290.00
    Log in for availability
    Each
    Each - $11,290.00
    $11,290.00
    Case of 4
       $11,290.00
    Each
       $11,290.00
    Supplier Number: 24013
  • HYPERStack® - 36 layer cell culture vessels, dual vent filter, Corning® CellBIND® surface
    3920 ml
    18000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $11,060.00
    $11,060.00
    Log in for availability
    Each
    Each - $11,060.00
    $11,060.00
    Case of 2
       $11,060.00
    Each
       $11,060.00
    Supplier Number: 24036
  • HYPERStack® - 36 layer cell culture vessels, dual vent filter, non-treated surface
    3920 ml
    18000 cm² (500 cm² per layer)
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $11,060.00
    $11,060.00
    Log in for availability
    Each
    Each - $11,060.00
    $11,060.00
    Case of 2
       $11,060.00
    Each
       $11,060.00
    Supplier Number: 24037