EasyReach™ Cleaning Tools, Contec® | Avantor
EasyReach™ Cleaning Tools, Contec®
EasyReach™ Cleaning Tools, Contec®
Supplier:  CONTEC, INC. MX
Certificates
EasyReach™ Cleaning Tools, Contec®
EasyReach™ Cleaning Tools, Contec®
8 Options Available Below
Product Details & Documents
The lightweight, stainless steel EasyReach™ Cleaning Tool is designed for cleaning isolators, laminar air flow hoods, glove boxes, and other mini environments. Rectangular shape makes overlapping straight-line action easy to achieve. Hinged joint and lightweight design makes one-handed cleaning comfortable and improves access to hard-to-reach areas.
  • Rounded tool edges and strong, durable, double-layer pads ensure equipment is not damaged during use
  • Lightweight EasyReach™ Tool can easily be manipulated, is comfortable to use, and minimizes fatigue
  • All components are designed to be chemical resistant, minimizing the risk of contamination from degradation
  • Presaturated option removes the need for 70% IPA spray bottles, reducing inventory and minimizing waste costs
  • Sorbent double layer Quiltec sealed edge polyester cleaning pads ensure particles and fibers are minimized
  • Polyester cleaning pads leave no unwanted surface residues
  • Available validated sterile
The single-use 100% polyester EasyReach™ cleaning pads provide a low level of particulates which is essential for use in product contact areas. The pads are very absorbent, ensuring even coverage of disinfectant or IPA. Easy grab edge on the cleaning pad ensures the pad fits tightly on the mop frame, but is easy to replace with gloved hands. Sterile pads presaturated with 70% IPA are also available.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Sterility
    Packed
    Availability
    Price
  • EasyReach™ 100% knitted polyester cleaning pads
    190×71 mm (7.5×2.8")
    Non sterile
    4/bag, 15 bags/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $611.46
    $611.46
    Prix promotionnel pour les nouveaux clients
    Case of 60
       $611.46
    Supplier Number: MEQT0001
  • EasyReach™ stainless steel handle and frame
    43.2 cm (17")
    Non sterile
    1/case
    Discontinued
    Supplier Number: -
  • EasyReach™ stainless steel mop frame
    19 cm (7.5")
    Non sterile
    1/case
    Discontinued
    Supplier Number: -
  • EasyReach™ sterile 100% knitted polyester cleaning pads
    190×71 mm (7.5×2.8")
    Sterile
    4/bag, 15 bags/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $1,720.00
    $1,720.00
    Case of 60
       $1,720.00
    Supplier Number: MEQT0002SZ
  • EasyReach™ sterile 100% knitted polyester cleaning pads
    6.9×19.6 cm (2.75×7.75")
    Sterile
    4/bag, 15 bags/case
    Discontinued
    Supplier Number: -
  • EasyReach™ stainless steel lay flat mop frame
    19 cm (7.5")
    Non sterile
    1/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $434.00
    $434.00
    Case of 1
       $434.00
    Supplier Number: 2685LF
  • EasyReach™ sterile 100% knitted polyester cleaning pads, presaturated with 70% IPA and 30% DI water
    190×71 mm (7.5×2.8")
    Sterile
    8/bag, 8 bags/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 64
    Case of 64 - $1,049.20
    $1,049.20
    Prix promotionnel pour les nouveaux clients
    Case of 64
       $1,049.20
    Supplier Number: PSME0001
  • EasyReach™ sterile ultrasonically sealed polyester cleaning pads
    190×71 mm (7.5×2.8")
    Sterile
    4/bag, 25 bags/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $1,006.20
    $1,006.20
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $1,006.20
    $1,006.20
    Prix promotionnel pour les nouveaux clients
    Case of 100
       $1,006.20
    La quantité minimale de commande est de 1
    Each
       $1,006.20
    La quantité minimale de commande est de 1
    Supplier Number: MEQT0022
  • EasyReach™ ultrasonically sealed polyester cleaning pads
    190×71 mm (7.5×2.8")
    Non sterile
    4/bag, 25 bags/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $808.40
    $808.40
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $808.40
    $808.40
    Prix promotionnel pour les nouveaux clients
    Case of 100
       $808.40
    La quantité minimale de commande est de 1
    Each
       $808.40
    La quantité minimale de commande est de 1
    Supplier Number: MEQT0021
  • EasyReach™ stainless steel one-piece lay flat frame
    42 cm (16.5")
    Non sterile
    1/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $448.00
    $448.00
    Case of 1
       $448.00
    Supplier Number: 2665LF
  • QuickConnect™ EasyReach™ polypropylene lay flat frame
    19 cm (7.5")
    Non sterile
    1/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $17.54
    $17.54
    Prix promotionnel pour les nouveaux clients
    Each
       $17.54
    Supplier Number: 2685P