Closures for RSA™ Screw-Thread Vials, Amber, MicroSolv
PP, Fitted Silicone Rubber/PTFE, ultra-pure, 9-425 mm, Light blue, Screw cap
97055-428
: 9509S-10C-B
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap
- Septum:Fitted Silicone Rubber/PTFE, ultra-pure
- Matière:PP
- Cap Color:Light blue
Accessories
- MaterialSeptumCap Size
- MaterialSeptumCap Size
- DescriptionCapacityMaximum Capacity
- 97051-398RSA-Pro™ low volume inserts, surface treated, with a conical, precision point and plastic spring attached. For use in 2 ml autosampler vials0.30 ml0.30 mlPack of 100 - $369.73
- DescriptionCapacityMaximum Capacity
- 97051-396RSA™ low volume inserts, precision point, with plastic spring attached. For use in 2 ml autosampler vials0.30 ml0.30 mlPack of 100 - $226.87