Nunc® Centrifuge Tubes, Polypropylene, Conical, Sterile, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
The Nunc® conical sterile polypropylene centrifuge tubes are premium, high quality, and environmentally friendly conical tubes.
- Plastic labware is a safer alternative to glass without sacrificing accuracy
These tubes have a high RCF rating that enables a greater range of applications, from low speed to super speed centrifugation. They are also USP Class VI, non-pyrogenic, non-cytotoxic and RNase/DNase free to ensure the highest performance.
Thermo Scientific Nunc® labware products are made from high purity resins, and molded using our state-of-the-art processes.
The tubes have leakproof construction and offer increased traceablilty with the largest writing area on the market. Additionally, they offer a reduced contamination risk with a recyclable plastic rack.
Ordering information: Tubes 89174-470 and 89174-474 includes reusable and recyclable polypropylene rack.
Frequently Bought Together