JACKSON SAFETY® V80 MONOGOGGLE™ 211 Protective Goggles, KIMBERLY-CLARK PROFESSIONAL®
Patented indirect ventilation system provides increased airflow, minimizes condensation and fogging, and protects against splash.
- Pliable plastic nosepiece for snug, comfortable seal
- Can be worn over most prescription eyeglasses
- Goggles provide 99.9% U.V. protection
Soft-frame design features a pantascopic angle to improve work area visibility.
: Meets ANSI Z87.1+ standards.
Frequently Bought Together