JACKSON SAFETY® V30 NEMESIS™ VL Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Fournisseur: Ansell Healthcare
Certificats
À propos de cet article
Sporty, frameless, lightweight design enhances comfort.
- Base curve 8
- Slip-stop temples to reduce slipping
- Compact profile accommodates smaller faces
No-brow design increases upward and peripheral vision.
Certifications: Eyewear meets ANSI Z87.1+ standards.
Delivery: Every pair includes a neck cord.
Frequently Bought Together