Corning® Gosselin™ Containers with Snap Caps, Corning
Fournisseur: Corning
Certificats
À propos de cet article
These containers are made of transparent or white PP.
- Lightweight and break-resistant
- Assembled or non-assembled
- Molded graduations
Ideal for sampling, storage and transport of liquid and solid samples.
Frequently Bought Together