WEBRIL™ Undercast Padding, Covidien
A 100% cotton fiber cushioning for synthetic or plastic casting material.
Padding minimizes dimensional changes and shifting under the cast, and will not collapse as moisture is absorbed.
Frequently Bought Together