ContactUsLinkComponent
2D-Xtract™, G-Biosciences
2D-Xtract™, G-Biosciences
Catalog # CA95043-430
Supplier:  G-Biosciences
CAS Number:  
undefined
2D-Xtract™, G-Biosciences
Catalog # CA95043-430
Supplier:  G-Biosciences
Supplier Number:  786-501
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    2D-Xtract™
  • Size:
    50 ml
  • Cat. no.:
    CA95043-430
  • Supplier No.:
    786-501
Specifications
About this item
2D-Xtract™ is a dry, urea-based and ready-to-use protein solubilization buffer.
  • Convenient and simple to use extraction buffer
  • No preparation required, simply hydrate and use
  • Prevents urea induced protein carbamylation
  • Prevents waste of unused reagents
  • No artifactual protein bands due to dust and human skin contamination
2D-Xtract™ is a proprietary modification of urea based chaotropic extraction buffers. Normal urea based buffers solubilize proteins effectively however can modify the native charge of the proteins, due to carbamylation. A process that modifies amino groups, inducing changes in the isoelectric point of proteinsleading to artifactual 2D results.
2D-Xtract™ is a dry, pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulfobetaine (ND SB 201) for maximumsolubilizing strength. The dry format allows researchers to freshly rehydrate as much or as little as is required and therefore prevent urea induced carbamylation.
This is suitable for sample extraction and solubilization for 2D gel electrophoresis and other applications and also suitable for rehydration of IPG Strips.
Ordering information: Accessories are available.
Caution: The kit is shipped at ambient temperature. Upon arrival, store it as specified on the bottle labels.