Operating Scissors, Lightweight, Integra™ Miltex® | Avantor
Operating Scissors, Lightweight, Integra™ Miltex®
Sharp/Blunt, 12.7 cm (5")
Catalog # 21909-290
Supplier:   Miltex
Fournisseur #:  5-61
Operating Scissors, Lightweight, Integra™ Miltex®
Catalog # 21909-290
Supplier:  Miltex
Operating Scissors, Lightweight, Integra™ Miltex®
Operating Scissors, Lightweight, Integra™ Miltex®
Sharp/Blunt, 12.7 cm (5")
Price
$224.17
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Delicate pattern designed for pediatric, vascular and other delicate procedures
Straight blades.
Spécifications
  • Désignation:
    Sharp/Blunt
  • Longueur:
    12.7 cm (5")