Operating Scissors, Integra™ Miltex®
 :   Miltex
Operating Scissors, Integra™ Miltex®
 :  Miltex
Operating Scissors, Integra™ Miltex®
Operating Scissors, Integra™ Miltex®
Standard pattern
Stainless steel
Product Family Options
  • Désignation
    Longueur
    Blade Type
  • Sharp/Sharp
    14.0 cm (5½”)
    Straight
    Each
    Each - $182.00
    $182.00
    Each
       $182.00
     : 5-6
  • Blunt/Blunt
    14.0 cm (5½”)
    Straight
    Each
    Each - $163.97
    $163.97
    Each
       $163.97
     : 5-26
  • Sharp/Blunt
    14.0 cm (5½”)
    Straight
    Each
    Each - $179.44
    $179.44
    Each
       $179.44
     : 5-16
  • Sharp/Sharp
    13.0 cm (5")
    Straight
    Each
    Each - $187.19
    $187.19
    Each
       $187.19
     : 5-4
  • Sharp/Sharp
    15.2 cm (6")
    Curved
    Each
    Each - $180.22
    $180.22
    Each
       $180.22
     : 5-37
  • Sharp/Blunt
    11.4 cm (4½")
    Curved
    Each
    Each - $194.99
    $194.99
    Each
       $194.99
     : 5-48
  • Sharp/Sharp
    15.2 cm (6")
    Straight
    Each
    Each - $208.54
    $208.54
    Each
       $208.54
     : 5-7
  • Sharp/Sharp
    14.0 cm (5½”)
    Curved
    Each
    Each - $181.14
    $181.14
    Each
       $181.14
     : 5-36
  • Blunt/Blunt
    14.0 cm (5½”)
    Curved
    Each
    Each - $168.27
    $168.27
    Each
       $168.27
     : 5-56
  • Sharp/Blunt
    15.2 cm (6")
    Straight
    Each
    Each - $180.47
    $180.47
    Each
       $180.47
     : 5-17
  • Sharp/Sharp
    11.4 cm (4½")
    Straight
    Each
    Each - $195.72
    $195.72
    Each
       $195.72
     : 5-2
  • Blunt/Blunt, Tungsten Carbide
    14.0 cm (5½”)
    Straight
    Each
    Each - $594.89
    $594.89
    Each
       $594.89
     : 5-26TC
  • Sharp/Sharp
    11.4 cm (4½")
    Curved
    Each
    Each - $201.77
    $201.77
    Each
       $201.77
     : 5-32
  • Sharp/Blunt
    13.0 cm (5")
    Straight
    Each
    Each - $143.14
    $143.14
    Each
       $143.14
     : 5-14
  • Sharp/Blunt
    11.4 cm (4½")
    Straight
    Each
    Each - $183.30
    $183.30
    Each
       $183.30
     : 5-12
  • Sharp/Blunt, Tungsten Carbide
    14.0 cm (5½”)
    Straight
    Each
    Each - $1,022.39
    $1,022.39
    Each
       $1,022.39
     : 5-16TC