Duramark® Lock Boxes
Supplier:   Therapak, LLC
Duramark® Lock Boxes
Supplier:  Therapak, LLC
Certificates
Duramark® Lock Boxes
8 Options Available Below
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Plastic lock box
    34.3×26×17.1 cm (13½×10¼× 6¾”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $450.61
    $450.61
    Each
       $450.61
    Supplier Number: 36745
  • Plastic lock box with over-door strap
    34.3×26×17.1 cm (13½×10¼× 6¾”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $445.13
    $445.13
    Each
       $445.13
    Supplier Number: 36506
  • Plastic lock box with under-door strap
    34.3×26×17.1 cm (13½×10¼× 6¾”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $549.42
    $549.42
    Each
       $549.42
    Supplier Number: 36743
  • Generic printed metal lock box with hinged over door strap with key lock
    35.5×26.7×17.8 cm (14×10½×7”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $424.55
    $424.55
    Each
       $424.55
    Supplier Number: 36507
  • Metal lock box
    35.5×26.7×17.8 cm (14×10½×7”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $647.03
    $647.03
    Each
       $647.03
    Supplier Number: 36746
  • Metal lock box with combination lock
    35.5×26.7×17.8 cm (14×10½×7”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $574.02
    $574.02
    Each
       $574.02
    Supplier Number: 36619
  • Metal lock box with combination lock and over-door strap
    35.5×26.7×17.8 cm (14×10½×7”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $597.42
    $597.42
    Each
       $597.42
    Supplier Number: 36620
  • Metal lock box with under-door strap
    35.5×26.7×17.8 cm (14×10½×7”)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $685.16
    $685.16
    Each
       $685.16
    Supplier Number: 36744
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Under-Door Strap for Metal or Plastic Box
    Under-Door Strap for Metal or Plastic Box
    89170-896
    Log in pour la disponibilité
    Each - $65.17
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Over-Door Strap for Metal or Plastic Box
    Over-Door Strap for Metal or Plastic Box
    89170-894
    Log in pour la disponibilité
    Each - $69.42
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Extra Long Over-Door Strap for Metal or Plastic Box
    Extra Long Over-Door Strap for Metal or Plastic Box
    10497-106
    Log in pour la disponibilité
    Each - $188.06
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Key for Metal or Plastic Box
    Replacement Key for Metal or Plastic Box
    89170-898
    Log in pour la disponibilité
    Each - $14.34