Camco® Wright Stain Buffer, pH 7.15, Cambridge
4.4 g
Catalog # CACMB41363
 :  402
Camco® Wright Stain Buffer, pH 7.15, Cambridge
Catalog # CACMB41363
CAS Number:  
Camco® Wright Stain Buffer, pH 7.15, Cambridge
Camco® Wright Stain Buffer, pH 7.15, Cambridge
4.4 g
CAS Number:  
Price
$448.31
Log in for availability
Pack of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each vial contains enough buffer to prepare one gallon of pH 7.15 buffer solution for use in the five-step staining process
The buffer is helpful with the development of the correct, clear nuclear color schemes.
Specifications
Specifications
  • Taille:
    4.4 g