To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Split Septum Device
- Cat. no.:CABD385100
- Supplier No.:385100
Split septum device is designed to reduce catheter-related bloodstream infections associated with needleless IV systems
The smooth surface is easy to clean because there are no crevices or gaps to house bacteria. The simple fluid path design reduces places for microbes to grow and delivers a better flow rate. Clear housing allows for visual assessment of the fluid path. The Acacia Extension Set includes a 15cm (6”) small bore bi-extension and spin nut.