Luer Tip Cap Tray, BD Medical
Catalog # CABD8341
Supplier: BD
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Tip Cap Tray
- Cat. no.:CABD8341
- Supplier No.:308341
Specifications
About this item
Latex-free caps are packaged for ease of application and minimized touch-contamination risk
The cover peels back to allow access to caps as needed, preserving sterility for remaining caps. Slip-on design allows for quick application. Self-vented tray packaging reduces waste. Tray contains ten caps.