Puritan® Cotton Tipped Applicators, Puritan Medical Products
Catalog # 89198-866
Supplier: Puritan Medical Products
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Cat. no.:89198-866
- Tip Size:17.4 mm (0.687")
- Handle Length:149.2 mm (5 9/16")
- Tip Diameter:6.35 mm (0.203")
- Overall Length:152.4 mm (6")
- Handle Color:Pink
- Handle Diameter:2.54 mm (0.1")
- Head Width:5.2 mm (7/32")
- Head Length:17.5 mm (¹¹⁄₁₆")
- Handle Width:2.5 mm (1/8")
- Supplier No.:25-806 2PC P
Specifications
About this item
These applicators feature high quality USP grade cotton that is bonded securely to a semi-flexible polystyrene handle by an aqueous based adhesive.
Absorbent tips are ideal for applying medicine, collecting cells, or cleaning wounds. Shorter swabs are suited for pediatric uses. Sterile. Available in three different handle colors (blue, pink, yellow). Made in USA.
Ordering information: Swabs are packaged two per envelope.