Puritan PurFlock Ultra® Flocked Swabs, Puritan Medical Products
Supplier: Puritan Medical Products
Certificates
About this item
Engineered for purity and maximal cell yield with a small sample size
The uniquely designed fiber structure accounts for better cell yield in the specimen where sample size is limited.
Standard tip swabs are ideal for Buccall cell collection. Mini tip and Ultrafine tip swabs are best suited for Nasopharyngeal and male urethral cell collection. Micro Ultrafine tip swabs are ideal for pediatric applications. All swabs are made in the USA.
PurFlock Ultra® specimen collection devices are ideal for PCR, molecular assays, DFA testing, forensic applications, and direct antigen testing.
Certifications: CE marking for medical use. Patented: United States, Canada, Europe, Australia, Japan, and China. Patents Pending: Hong Kong.
Ordering information: Sterile swabs are individually wrapped. Nonsterile swabs are bulk packaged.
Frequently Bought Together