TRACK-IT™ AC Event Logger, Monarch Instrument | Avantor
TRACK-IT™ AC Event Logger, Monarch Instrument
Supplier:   Monarch Instrument
TRACK-IT™ AC Event Logger, Monarch Instrument
Supplier:  Monarch Instrument
Certificates
TRACK-IT™ AC Event Logger, Monarch Instrument
2 Options Available Below
Product Details & Documents
The AC event data logger is a battery powered stand alone compact data logger that records up to 64000 on/off cycles.
Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours or can be set to store a sample at every event. The on board data storage is non‐volatile so data will not be lost in the event of a depleted battery.
The Track‐It™ AC event logger can also be used to record ambient temperature along with on/off cycles. This can help diagnose heat vs. cycle time issues. Information displayed on the LCD is also user programmable and can be sequenced by pressing the logger button. The multicolor LED indicator notifies the user of alarms, or general activity.
Use the Track‐It™ AC event logger to monitor cycle patterns of pumps, blowers and other electrical equipment powered by 120 or 240 Vac. The unit is easily configured using the included Track‐It™ software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger.
Ordering information: Interactive Software included.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89184-570
  • 89184-572
  • Alarm:
  • High/low level alarms
  • High/low level alarms
  • Display:
  • LCD, 4 digit
  • LCD, 4 digit
  • Interface:
  • USB
  • USB
  • Battery life:
  • 1 year (1 min sampling rate)
  • 3 years (1 min sampling rate)
  • W×D×H:
  • 25.4×103.2×16.7 mm
  • 25.4×103.2×16.7 mm
  • Data memory - No. of readings:
  • 64000
  • 64000
  • Voltage:
  • 120 - 240 V
  • 120 - 240 V
  • Batteries:
  • Lithium CR2032 battery
  • Lithium EF65162 battery
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Résolution
    Accuracy
    Availability
    Price
  • TRACK-IT™ AC Event Logger with LCD display and standard battery
    T: −20...+70 °C
    T: 0.06 °C
    T: ±0.5 °C (0...50 °C), ±2 °C (−20...85 °C)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $406.24
    $406.24
    Each
       $406.24
    Supplier Number: 5396-0711
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89184-570
    Alarm
    High/low level alarms
    Display
    LCD, 4 digit
    Interface
    USB
    Battery life
    1 year (1 min sampling rate)
    W×D×H
    25.4×103.2×16.7 mm
    Data memory - No. of readings
    64000
    Voltage
    120 - 240 V
    Batteries
    Lithium CR2032 battery
    Small Business Enterprise
  • TRACK-IT™ AC Event Logger with LCD display and long life battery
    T: −20...+85 °C
    T: 0.06 °C
    T: ±0.5 °C (0...50 °C), ±2 °C (−20...85 °C)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $482.63
    $482.63
    Each
       $482.63
    Supplier Number: 5396-0712
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89184-572
    Alarm
    High/low level alarms
    Display
    LCD, 4 digit
    Interface
    USB
    Battery life
    3 years (1 min sampling rate)
    W×D×H
    25.4×103.2×16.7 mm
    Data memory - No. of readings
    64000
    Voltage
    120 - 240 V
    Batteries
    Lithium EF65162 battery
    Small Business Enterprise
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • 3 ft USB 2.0 interface cable
    3 ft USB 2.0 interface cable
    89184-592
    Log in pour la disponibilité
    Each - $29.86
Informations sur le produit
  • Catalog #
    Availability
    Price
  • TRACK-IT™ software replacement CD
    TRACK-IT™ software replacement CD
    89184-594
    Log in pour la disponibilité
    Each - $74.68
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Standard Battery: Coin cell lithium CR2032
    Replacement Standard Battery: Coin cell lithium CR2032
    89184-598
    Log in pour la disponibilité
    Each - $40.50
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Replacement Long Life Battery: EF651625
    Replacement Long Life Battery: EF651625
    89184-600
    Log in pour la disponibilité
    Each - $63.67