Cogent UDC-Cholesterol™ TYPE-C Silica™ HPLC Columns, MicroSolv
Supplier: MicroSolv Technology Corp
Certificates
About this item
These mid range columns work in three chromatographic modes: reverse phase, normal phase, and aqueous normal phase.
Their silicon-carbon bonds make these columns hydrolytically stable as a stationary phase. They will produce unique selectivity for many compounds, especially at lower temperatures, and can separate some compounds based on molecular shape.
Ordering information: Guard column kits contain five guard columns with or without a holder.
Frequently Bought Together