Suite of Columns, Method Development Kits, MicroSolv
Suite of Columns, Method Development Kits, MicroSolv
Suite of Columns, Method Development Kits, MicroSolv
Certificates
Suite of Columns, Method Development Kits, MicroSolv
Suite of Columns, Method Development Kits, MicroSolv
11 Options Available Below
Product Details & Documents
Complete set of columns for method developers that need and want the full range of selectivities on hand at all times.
Each set contains one each of Bidentate C18™, UDC-Cholesterol™, Bidentate C8™, Diamond-Hydride™ and Silica-C™ columns in the same configuration.
Range from extremely hydrophobic to extremely hydrophilic, from low pH to high pH, from reversed phase to aqueous normal phase to normal phase with only five columns.
Ordering information: Guard column suites contain five guard columns with or without a holder.
Product Family Options
Columns
Informations sur le produit
  • Désignation
    Type de colonne
    Taille de pore
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • 5 Column Suite
    Capillaire
    100 Å
    4 µm
    1.00 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,250.52
    $10,250.52
    Each
       $10,250.52
    Minimum order quantity is 1
    Supplier Number: 40100-15P-1
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    2.10 mm
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,747.95
    $7,747.95
    Each
       $7,747.95
    Minimum order quantity is 1
    Supplier Number: 40100-05P-2
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    2.10 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,187.85
    $8,187.85
    Each
       $8,187.85
    Minimum order quantity is 1
    Supplier Number: 40100-7.5P-2
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    2.10 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,465.71
    $9,465.71
    Each
       $9,465.71
    Minimum order quantity is 1
    Supplier Number: 40100-10P-2
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    2.10 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,609.76
    $10,609.76
    Each
       $10,609.76
    Minimum order quantity is 1
    Supplier Number: 40100-15P-2
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    3.00 mm
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,747.95
    $7,747.95
    Each
       $7,747.95
    Minimum order quantity is 1
    Supplier Number: 40100-05P-3
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    4.60 mm
    50 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,980.45
    $7,980.45
    Each
       $7,980.45
    Minimum order quantity is 1
    Supplier Number: 40100-05P
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    4.60 mm
    75 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,433.52
    $8,433.52
    Each
       $8,433.52
    Minimum order quantity is 1
    Supplier Number: 40100-7.5P
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    4.60 mm
    100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,235.10
    $9,235.10
    Each
       $9,235.10
    Minimum order quantity is 1
    Supplier Number: 40100-10P
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    4.60 mm
    150 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10,558.00
    $10,558.00
    Each
       $10,558.00
    Minimum order quantity is 1
    Supplier Number: 40100-15P
  • 5 Column Suite
    Analytique
    100 Å
    4 µm
    4.60 mm
    250 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11,233.48
    $11,233.48
    Each
       $11,233.48
    Minimum order quantity is 1
    Supplier Number: 40100-25P