Metabolomics Column Kits, MicroSolv
Metabolomics Column Kits, MicroSolv
Certificates
Metabolomics Column Kits, MicroSolv
Metabolomics Column Kits, MicroSolv
4 Options Available Below
Product Details & Documents
Each kit includes one Cogent Bidentate C18™ column and one Cogent Diamond Hydride™ column.
Using these two columns in an LCMS method and the appropriate mass spectrometer, it is easy to create a reliable 'metabolome', which consists of plotting metabolite retention times versus their accurate masses. Using these metabolomes, one can find the differences between sample sets such as healthy cells and diseased cells.
The C18 column is used for the most hydrophobic compounds while the Diamond Hydride™ column is used for all other compounds.
Product Family Options
Product Information
  • Type de colonne
    Taille de pore
    Granulométrie
    Ø int.
    Longueur
    Availability
    Price
  • Capillaire
    100 Å
    4 µm
    1.00 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $5,842.49
    $5,842.49
    Each
       $5,842.49
    Supplier Number: 43000-10P-1
  • Analytique
    100 Å
    4 µm
    2.10 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $2,736.48
    $2,736.48
    Each
       $2,736.48
    Supplier Number: 43000-05P-2
  • Analytique
    100 Å
    4 µm
    2.10 mm
    100 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,241.99
    $3,241.99
    Each
       $3,241.99
    Supplier Number: 43000-10P-2
  • Analytique
    100 Å
    4 µm
    2.10 mm
    150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,665.60
    $3,665.60
    Each
       $3,665.60
    Supplier Number: 43000-15P-2