Cogent Bidentate C8™ TYPE-C Silica™ HPLC Columns, MicroSolv
Using proprietary bonding technology, columns are bonded directly to the nonpolar TYPE-C™ silica surface with two separate points of attachment, allowing them to operate in three different domains: reverse phase, normal phase and aqueous normal phase.
: Guard column kits contain five guard columns with or without a holder.
Frequently Bought Together