HISTOSETTE® I Tissue Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® I Tissue Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® I Tissue Processing and Embedding Cassettes, Simport Scientific
Supplier:  Simport Scientific
Certificates
HISTOSETTE® I Tissue Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® I Tissue Processing and Embedding Cassettes, Simport Scientific
14 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lids snap apart and lock easily into the base of the cassettes. The anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments.
Ordering information: Packed in three dispenser boxes of 500 cassettes. Fluorescent colored cassettes require a minimum order quantity of 40 cases for purchase.
Product Family Options
Informations sur le produit
  • Color
    Availability
    Price
  • Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-3
  • Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-4
  • Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-5
  • Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-6
  • Peach
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-7
  • Tan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-8
  • Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-9
  • Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-11
  • Aqua
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-12
  • Fluorescent Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-16
  • Fluorescent Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-17
  • Fluorescent Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $370.57
    $370.57
    Case of 1,500
       $370.57
    Supplier Number: M498-18
  • White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $531.03
    $531.03
    Case of 1,500
       $531.03
    Supplier Number: M498-2
  • Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $437.02
    $437.02
    Case of 1,500
       $437.02
    Supplier Number: M498-10