HISTOSETTE® II Tissue Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® II Tissue Processing and Embedding Cassettes, Simport Scientific
Supplier:  Simport Scientific
Certificates
HISTOSETTE® II Tissue Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® II Tissue Processing and Embedding Cassettes, Simport Scientific
11 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable cassettes hold tissue specimens efficiently during the embedding process and in storage
Molded from acetal polymer, they keep specimens submerged in liquid and are resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. Anterior writing area is angled at 45° for suitability with certain types of cassette labeling instruments. Colors are non-cytotoxic and nonmetallic
 : Lids are supplied unattached. Packed in two dispenser boxes of 500 cassettes and one dispenser box of 1000 lids.
Product Family Options
Informations sur le produit
  • Color
    Availability
    Price
  • White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $343.64
    $343.64
    Case of 1,000
       $343.64
    Supplier Number: M485-2
  • Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $345.93
    $345.93
    Case of 1,000
       $345.93
    Supplier Number: M485-3
  • Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $341.97
    $341.97
    Case of 1,000
       $341.97
    Supplier Number: M485-5
  • Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $345.93
    $345.93
    Case of 1,000
       $345.93
    Supplier Number: M485-6
  • Peach
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $345.93
    $345.93
    Case of 1,000
       $345.93
    Supplier Number: M485-7
  • Tan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $254.13
    $254.13
    Case of 1,000
       $254.13
    Supplier Number: M485-8
  • Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $345.93
    $345.93
    Case of 1,000
       $345.93
    Supplier Number: M485-9
  • Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $256.49
    $256.49
    Case of 1,000
       $256.49
    Supplier Number: M485-10
  • Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $256.49
    $256.49
    Case of 1,000
       $256.49
    Supplier Number: M485-11
  • Aqua
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $256.49
    $256.49
    Case of 1,000
       $256.49
    Supplier Number: M485-12
  • Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $341.97
    $341.97
    Case of 1,000
       $341.97
    Supplier Number: M485-4