MICROSETTE™ I Biopsy Processing and Embedding Cassettes, Simport Scientific
À propos de cet article
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
: Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.
13 Options Available Below
Product Details & Documents
These innovative disposable plastic cassettes eliminate the need for biopsy pads
Cover and base feature over 2000 square openings, each with a diameter of 0.26mm. These mesh holes allow for maximum fluid exchange. The high-density acetal polymer is resistant to the chemical action of histological solvents. The integral one-piece lids can be opened and closed multiple times. The anterior writing area is set at a 45° angle. Compatible with most cassette printers.
: Packaged in dispenser boxes of 150 cassettes with covers assembled, four boxes per case.
Product Family Options
Informations sur le produit
- ColorAvailabilityPrice
- WhiteDiscontinued-Supplier Number: M502-2
- PinkPack of 250$251.31Case of 1000$1,005.24Supplier Number: M502-3
- GreenPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-4
- YellowPack of 250$251.31Case of 1000$1,005.24Supplier Number: M502-5
- BluePack of 250$251.31Case of 1000$1,005.24Supplier Number: M502-6
- PeachPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-7
- TanPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-8
- GrayPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-9
- LilacPack of 250$162.81Case of 1000$651.24Supplier Number: M502-10
- OrangePack of 250$251.31Case of 1000$1,005.24Supplier Number: M502-11
- AquaPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-12
- Fluorescent PinkPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-16
- Fluorescent GreenPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-17
- Fluorescent YellowPack of 250$265.52Case of 1000$1,062.08Supplier Number: M502-18