SlideFolder™ Microscope Slide Holders, Simport Scientific
Fournisseur: Simport Scientific
Certificats
À propos de cet article
These two-part folders hold up to twenty standard microscope slides
- All units are stackable
- Not autoclavable
The high-impact polystyrene base holds slides horizontally with numbered spaces for easy identification. The transparent polystyrene door covers the slides and can be swung behind the folder for space-saving purposes.
Temperature range: –80 to 80°C (–112 to 176°F)
Frequently Bought Together