SlideFile™ Jr. Storage System, Simport Scientific
Holds up to 200 slides per unit and stackable for space-efficient storage
- Base, cover, and trays are made of high impact polystyrene
- Not autoclavable
The slide box includes a removable tray that holds the slides upright. For maximum storage space, remove the tray and line the slides in three rows. A tinted hinged cover makes the contents of the box easy to see.
Temperature range: –80 to 80°C (–112° to 176°F)
Frequently Bought Together