SLIMSETTE™ Tissue Processing and Embedding Cassettes, Simport Scientific
SLIMSETTE™ Tissue Processing and Embedding Cassettes, Simport Scientific
Supplier:  Simport Scientific
Certificates
SLIMSETTE™ Tissue Processing and Embedding Cassettes, Simport Scientific
SLIMSETTE™ Tissue Processing and Embedding Cassettes, Simport Scientific
14 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These compact, easy-to-use cassettes incorporate a unique recessed cover which allows more cassettes to be stacked
They are molded from acetal polymer to keep specimens safely submerged in liquid and are resistant to chemical actions of histological solvents. The 1x 5mm slotted openings ensure efficient fluid exchange and drainage, and the one-piece integral lid can be opened and closed as often as necessary. The anterior writing area is slanted at a 45° angle
 : CE marked.
 : Packaged in three dispenser boxes of 500 cassettes with covers assembled.
Product Family Options
Informations sur le produit
  • Color
    Availability
    Price
  • White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-2
  • Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-3
  • Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-4
  • Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-5
  • Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-6
  • Peach
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-7
  • Tan
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-8
  • Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-9
  • Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-10
  • Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-11
  • Aqua
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.64
    $493.64
    Case of 1,500
       $493.64
    Supplier Number: M509-12
  • Fluorescent Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.63
    $493.63
    Case of 1,500
       $493.63
    Supplier Number: M509-16
  • Fluorescent Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.63
    $493.63
    Case of 1,500
       $493.63
    Supplier Number: M509-17
  • Fluorescent Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,500
    Case of 1,500 - $493.63
    $493.63
    Case of 1,500
       $493.63
    Supplier Number: M509-18