UniMailer™ Slide Mailers, Simport Scientific
Fournisseur: Simport Scientific
Certificats
À propos de cet article
These one-slide trays are stackable and are secured by a unique locking mechanism
They are designed to accommodate standard slides with or without cover glasses. Mailers hold slides horizontally and are designed for an exact fit to prevent breaking, but slides are easily removed as needed
Temperature range: –80 to 80°C
Frequently Bought Together