Q700 Sonicator without a Probe, Qsonica
Catalog # 89184-808
Supplier: QSonica, LLC.
Specifications
- Description:Q700 Sonicator system (power supply, converter, cables and wrench set) excludes the standard probe/horn
- Cat. no.:89184-808
- Voltage:110 V
- Supplier No.:Q700A-110
Specifications
About this item
Q700 Sonicator system without a probe
Probe/Cup Horn must be ordered separately.
Accessories information: Horn extenders, also available in solid or tapped varieties, can reach into long neck vessels. Half-wave extenders add 12.7 cm (5") to the horn’s length; full-wave extenders add 25.4 cm (10") to the horn’s length.
Delivery information: Supplied with power supply, converter, cables and wrench set.