ClipTip® Empty Racks, Thermo Scientific
89173-428
- Description:ClipTip® Empty Rack, Small (10 x Rack)
- Cat. no.:89173-428
- Supplier No.:94410519
Empty racks for ClipTip® reload systems.
The small rack is compatible with ClipTip® 20 and 50 µl pipette tips.
: Manufactured according to ISO 9001 and ISO 13485. The F1-ClipTip® pipetting system is CE-marked in accordance with IVD Directive in Europe.