VWR® Pour Spout Sample Cups | Avantor
VWR® Pour Spout Sample Cups
192 ml (6.5 oz.), 72 mm (2 13/16"), 87 mm (3⁷⁄₁₆")
Catalog # 95042-200
Supplier:   VWR International
VWR® Pour Spout Sample Cups
Catalog # 95042-200
Supplier:  VWR International
VWR® Pour Spout Sample Cups
192 ml (6.5 oz.), 72 mm (2 13/16"), 87 mm (3⁷⁄₁₆")
Price
$386.46
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 500
Product Details & Documents
These translucent polystyrene containers are appropriate for use as urine cups or many other applications.
  • Polystyrene construction
  • Translucent siding
  • Suitable for a range of applications
They are graduated in ounces, cc, and mililiters, and have convenient pour spouts. They are packaged in sleeves of 25.
Spécifications
  • Ø:
    72 mm (2 13/16")
  • Capacité:
    192 ml (6.5 oz.)
  • Hauteur:
    87 mm (3⁷⁄₁₆")